Search Thermo Fisher Scientific
Gene Symbol: LINC00663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026956.1 | 3 | 834 | |||
GenBank mRNA | AK022418.1 | - | 1 | 468 | ||
AK289657.1 | - | 3 | 814 | |||
BC034236.1 | - | 4 | 966 |