Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000816.3 | NP_000807.2 | 6 | 1073 | ||
NM_198903.2 | NP_944493.2 | 7 | 1193 | |||
NM_198904.2 | NP_944494.1 | 6 | 1073 | |||
GenBank mRNA | AK299640.1 | - | 7 | 798 | ||
AK315200.1 | - | 6 | 725 | |||
BC059389.1 | - | 6 | 894 | 5288256 | ||
BC069348.1 | - | 6 | 809 | 7262321 | ||
BC074795.2 | - | 6 | 747 | 30915258 | ||
CD013896.1 | - | 6 | 851 | |||
CD013897.1 | - | 6 | 783 | |||
X15376.1 | - | 6 | 941 |