Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026834.1 | 2 | 1353 | |||
GenBank mRNA | BC033355.1 | - | 2 | 1344 | ||
BC039347.1 | - | 2 | 819 | |||
BC045572.1 | - | 2 | 1329 | |||
BC121796.1 | - | 1 | 419 |