Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 18 | 2004 | ||
XM_011527756.2 | XP_011526058.1 | 20 | 2109 | |||
XM_011527757.2 | XP_011526059.1 | 18 | 2162 | |||
XM_011527758.2 | XP_011526060.1 | 19 | 2022 | |||
XM_011527761.1 | XP_011526063.1 | 21 | 2179 | |||
XM_011527764.2 | XP_011526066.1 | 17 | 2075 | |||
XM_011527765.2 | XP_011526067.1 | 18 | 1923 | |||
XM_011527768.2 | XP_011526070.1 | 6 | 880 | |||
XM_017026438.1 | XP_016881927.1 | 21 | 2170 | |||
XM_017026439.1 | XP_016881928.1 | 20 | 2092 | |||
XM_017026440.1 | XP_016881929.1 | 19 | 2065 | |||
XM_017026441.1 | XP_016881930.1 | 9 | 1207 | |||
GenBank mRNA | AB075861.1 | - | 16 | 1701 | ||
BC028236.1 | - | 16 | 1646 |