Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145112.1 | NP_001138584.1 | 15 | 1675 | ||
XM_011521336.2 | XP_011519638.2 | 14 | 2403 | |||
XM_011521337.2 | XP_011519639.2 | 14 | 2538 | |||
XM_011521338.2 | XP_011519640.1 | 16 | 2102 | |||
XM_011521339.2 | XP_011519641.1 | 15 | 1983 | |||
XM_011521340.2 | XP_011519642.1 | 14 | 1924 | |||
XM_011521341.1 | XP_011519643.1 | 14 | 1687 | |||
XM_011521342.2 | XP_011519644.1 | 12 | 2584 | |||
XM_011521343.2 | XP_011519645.1 | 15 | 1934 | |||
XM_011521344.2 | XP_011519646.1 | 12 | 1376 | |||
XM_011521345.2 | XP_011519647.1 | 15 | 1896 | |||
XM_011521346.2 | XP_011519648.2 | 10 | 2110 | |||
XM_011521347.1 | XP_011519649.1 | 13 | 1492 | |||
XM_011521348.2 | XP_011519650.1 | 14 | 1590 | |||
XM_017022000.1 | XP_016877489.1 | 15 | 2493 | |||
XM_017022001.1 | XP_016877490.1 | 12 | 1524 |