Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523379.1 | XP_011521681.1 | 1 | 1014 | ||
XM_011523381.1 | XP_011521683.1 | 1 | 1014 | |||
GenBank mRNA | AK130680.1 | - | 1 | 927 |