Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039360.2 | NP_001034449.1 | 2 | 890 | ||
NM_001318841.1 | NP_001305770.1 | 4 | 1071 | |||
XM_005258229.4 | XP_005258286.1 | 3 | 815 | |||
XM_011525861.2 | XP_011524163.1 | 6 | 1295 | |||
XM_011525863.2 | XP_011524165.1 | 5 | 1412 | |||
XM_011525864.2 | XP_011524166.1 | 5 | 1167 | |||
XM_011525865.2 | XP_011524167.1 | 5 | 1408 | |||
XM_011525866.2 | XP_011524168.1 | 4 | 1153 | |||
XM_011525867.2 | XP_011524169.1 | 4 | 942 | |||
XM_011525869.2 | XP_011524171.1 | 5 | 1417 | |||
XM_011525870.2 | XP_011524172.1 | 4 | 1113 | |||
XM_011525871.2 | XP_011524173.1 | 2 | 1628 | |||
XM_017025605.1 | XP_016881094.1 | 6 | 3090 | |||
XM_017025606.1 | XP_016881095.1 | 6 | 1544 | |||
XM_017025607.1 | XP_016881096.1 | 6 | 1299 | |||
XM_017025608.1 | XP_016881097.1 | 5 | 2967 | |||
XM_017025609.1 | XP_016881098.1 | 4 | 1284 | |||
GenBank mRNA | AK296709.1 | - | 2 | 890 | ||
AM988836.1 | - | 2 | 878 | |||
AM988837.1 | - | 2 | 1023 | |||
AM988838.1 | - | 3 | 1073 | |||
AM988839.1 | - | 4 | 1071 | |||
EF120357.1 | - | 2 | 942 |