Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039396.1 | NP_001034485.1 | 1 | 683 | ||
GenBank mRNA | AK074166.1 | - | 1 | 613 | ||
AK097881.1 | - | 1 | 580 | |||
BC104997.1 | - | 1 | 548 | 8144000 | ||
BC112230.1 | - | 1 | 548 | 8327698 |