Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018593.4 | NP_061063.2 | 4, 5 | 1257 | ||
XM_017010237.1 | XP_016865726.1 | 3, 4 | 1363 | |||
GenBank mRNA | AB057445.1 | - | 4, 5 | 1257 | ||
AF116652.2 | - | 3, 4 | 204 | |||
AK126183.1 | - | 4, 5 | 1332 | |||
AK172789.1 | - | 2, 3 | 478 | |||
BC017968.1 | - | 2, 3 | 266 | |||
BC066985.1 | - | 4, 5 | 1229 | 4827423 |