Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015021.1 | NP_055836.1 | 3, 4 | 451 | ||
XM_005248697.4 | XP_005248754.1 | 3, 4 | 265 | |||
XM_011535625.2 | XP_011533927.1 | 3, 4 | 413 | |||
XM_017010575.1 | XP_016866064.1 | 4, 5 | 3642 | |||
XM_017010576.1 | XP_016866065.1 | 5, 6 | 1168 | |||
XM_017010577.1 | XP_016866066.1 | 3, 4 | 3497 | |||
XM_017010578.1 | XP_016866067.1 | 5, 6 | 6150 | |||
XM_017010579.1 | XP_016866068.1 | 5, 6 | 3176 | |||
XM_017010580.1 | XP_016866069.1 | 5, 6 | 597 | |||
XM_017010581.1 | XP_016866070.1 | 4, 5 | 583 | |||
XM_017010582.1 | XP_016866071.1 | 3, 4 | 3293 | |||
XM_017010583.1 | XP_016866072.1 | 4, 5 | 890 | |||
XM_017010584.1 | XP_016866073.1 | 3, 4 | 5266 | |||
GenBank mRNA | AK123473.1 | - | 3, 4 | 437 | ||
BX537746.1 | - | 3, 4 | 426 |