Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015188.1 | NP_056003.1 | 5 | 1529 | ||
XM_006717732.3 | XP_006717795.2 | 5 | 1529 | |||
XM_006717733.2 | XP_006717796.1 | 6 | 489 | |||
XM_011539558.2 | XP_011537860.1 | 5 | 540 | |||
XM_017015997.1 | XP_016871486.1 | 5 | 1529 | |||
XM_017015998.1 | XP_016871487.1 | 4 | 316 | |||
XM_017015999.1 | XP_016871488.1 | 4 | 532 | |||
GenBank mRNA | AB011180.1 | - | 5 | 1345 | ||
AB449898.1 | - | 5 | 1326 |