Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: THSD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015204.2 | NP_056019.1 | 3 | 1463 | ||
XM_006715659.1 | XP_006715722.1 | 3 | 1462 | |||
XM_006715660.1 | XP_006715723.1 | 3 | 1462 | |||
XM_006715662.1 | XP_006715725.1 | 3 | 1462 | |||
XM_011515193.2 | XP_011513495.1 | 3 | 3787 | |||
XM_011515194.1 | XP_011513496.1 | 3 | 1228 | |||
GenBank mRNA | AB023177.2 | - | 2 | 747 | ||
AK092252.1 | - | 3 | 1463 |