Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243794.1 | NP_001230723.1 | 2 | 1835 | ||
NM_001243795.1 | NP_001230724.1 | 2 | 1849 | |||
NM_018641.4 | NP_061111.1 | 2 | 1863 | |||
GenBank mRNA | AF239822.1 | - | 2 | 1774 | ||
BC002918.1 | - | 2 | 1824 | 3955396 | ||
BC015954.1 | - | 2 | 1824 |