Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018659.2 | NP_061129.1 | 4 | 720 | ||
XM_017008299.1 | XP_016863788.1 | 5 | 835 | |||
GenBank mRNA | AF193766.1 | - | 4 | 723 | ||
AF274956.1 | - | 4 | 580 | |||
AY359101.1 | - | 4 | 721 | |||
BC031391.1 | - | 4 | 696 | 5186334 |