Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099456.2 | NP_001092926.2 | 2 | 989 | ||
GenBank mRNA | AL365519.1 | - | 2 | 307 | ||
BC171886.1 | - | 1 | 1426 |