Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C2orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143959.1 | NP_001137431.1 | 4 | 518 | ||
NM_001143960.2 | NP_001137432.1 | 4 | 531 | |||
NM_001316317.1 | NP_001303246.1 | 3 | 417 | |||
XM_017003967.1 | XP_016859456.1 | 4 | 519 | |||
GenBank mRNA | BP422302.1 | - | 3 | 338 |