Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098844.2 | NP_001092314.1 | 4 | 1056 | ||
XM_011519627.2 | XP_011517929.1 | 4 | 933 | |||
XM_011519628.1 | XP_011517930.1 | 3 | 774 | |||
XM_011519629.1 | XP_011517931.1 | 3 | 822 | |||
XM_017016574.1 | XP_016872063.1 | 4 | 1782 | |||
GenBank mRNA | BC157856.1 | - | 4 | 1056 | 9057075 |