Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145010.1 | NP_001138482.1 | 9 | 1174 | ||
XM_011520634.2 | XP_011518936.1 | 9 | 1352 | |||
XM_011520635.2 | XP_011518937.1 | 7 | 851 | |||
XM_017019254.1 | XP_016874743.1 | 7 | 1108 | |||
XM_017019255.1 | XP_016874744.1 | 6 | 958 |