Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102723407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546198.1 | XP_011544500.1 | 2 | 358 | ||
GenBank mRNA | AF103061.2 | - | 1 | 206 | ||
AF103313.1 | - | 1 | 206 | |||
AF174124.1 | - | 239 | ||||
AF396470.1 | - | 1 | 239 | |||
AF396478.1 | - | 1 | 239 | |||
AF455548.1 | - | 1 | 239 | |||
AJ244933.1 | - | 1 | 239 | |||
AJ244980.1 | - | 1 | 239 | |||
AJ389176.1 | - | 1 | 239 | |||
AJ414019.1 | - | 1 | 239 | |||
AJ579202.1 | - | 4 | 242 | |||
AY393167.1 | - | 1 | 192 | |||
AY671339.1 | - | 1 | 232 | |||
DQ100904.1 | - | 1 | 235 | |||
DQ454459.1 | - | 2 | 152 | |||
DQ454670.1 | - | 1 | 164 | |||
DQ454700.1 | - | 1 | 152 | |||
DQ454827.1 | - | 1 | 161 | |||
DQ454834.1 | - | 1 | 155 | |||
DQ454952.1 | - | 1 | 155 | |||
DQ523952.1 | - | 1 | 236 | |||
DQ523972.1 | - | 236 | ||||
DQ523999.1 | - | 1 | 236 | |||
DQ524000.1 | - | 1 | 236 | |||
EF175423.1 | - | 174 | ||||
EF175437.1 | - | 174 | ||||
EU099126.1 | - | 1 | 188 | |||
FJ039783.1 | - | 1 | 239 | |||
HM152512.1 | - | 1 | 239 | |||
HM152518.1 | - | 2 | 242 | |||
HM996243.1 | - | 1 | 241 | |||
JN376176.1 | - | 1 | 164 | |||
JN376188.1 | - | 1 | 164 | |||
JN376230.1 | - | 1 | 164 | |||
JN376232.1 | - | 1 | 164 | |||
JN376233.1 | - | 1 | 164 | |||
JN376234.1 | - | 1 | 164 | |||
JN376235.1 | - | 1 | 164 | |||
JN376309.1 | - | 1 | 164 | |||
JN376492.1 | - | 1 | 160 | |||
KU674890.1 | - | 1 | 192 | |||
S45207.1 | - | 404 | ||||
S45947.1 | - | 296 | ||||
Z14195.1 | - | 2 | 333 |
Gene Symbol: IGHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063384.1 | - | 2 | 317 |
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC011857.2 | - | 3 | 315 |
Gene Symbol: IGHV4-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC073766.1 | - | 3 | 313 | 4855124 |
Gene Symbol: LOC102724971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543804.2 | XP_011542106.1 | 2 | 413 |
Gene Symbol: LOC107983983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022769.1 | XP_016878258.1 | 2 | 419 |
Gene Symbol: SKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF174049.1 | - | 1 | 236 |