Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000365.5 | NP_000356.1 | 7 | 1226 | ||
NM_001159287.1 | NP_001152759.1 | 7 | 1335 | |||
NM_001258026.1 | NP_001244955.1 | 7 | 1469 | |||
GenBank mRNA | AK222638.1 | - | 7 | 1215 | ||
BC017165.1 | - | 6 | 972 |