Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL6 SIRNA Assays ›
Gene Symbol:
BCL6
Gene Name:
B-cell CLL/lymphoma 6
Gene Aliases:
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
Chromosome Location:
Chr.3: 187721377 - 187745725 on Build GRCh38
UniGene ID:
Hs.478588
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
275043
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005247694.3
XP_005247751.1
1
305
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
negative regulation of cell-matrix adhesion
germinal center formation
regulation of germinal center formation
negative regulation of B cell apoptotic process
transcription, DNA-templated
inflammatory response
cellular response to DNA damage stimulus
Rho protein signal transduction
spermatogenesis
negative regulation of cell proliferation
actin cytoskeleton organization
myeloid cell differentiation
B cell differentiation
negative regulation of cell growth
positive regulation of B cell proliferation
positive regulation of histone deacetylation
negative regulation of mast cell cytokine production
negative regulation of Rho protein signal transduction
type 2 immune response
regulation of cell proliferation
regulation of apoptotic process
positive regulation of apoptotic process
regulation of GTPase activity
regulation of memory T cell differentiation
negative regulation of T-helper 2 cell differentiation
positive regulation of neuron differentiation
negative regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
negative regulation of isotype switching to IgE isotypes
erythrocyte development
regulation of inflammatory response
regulation of neurogenesis
regulation of immune response
positive regulation of cellular component movement
negative regulation of cellular senescence
RNA polymerase II regulatory region sequence-specific DNA binding
intronic transcription regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
chromatin DNA binding
sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0