Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125823.1 | 2 | 376 | |||
GenBank mRNA | BX094193.1 | - | 2 | 148 | ||
DB446853.1 | - | 2 | 328 |