Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018929.2 | NP_061752.1 | 1 | 368 | ||
NM_032407.1 | NP_115783.1 | 1 | 368 | |||
GenBank mRNA | AF152339.1 | - | 1 | 368 | ||
AF152526.1 | - | 1 | 368 | |||
BC112081.1 | - | 1 | 442 | 8327549 | ||
BC112083.1 | - | 1 | 442 | 8327551 |