Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001895.2 | NP_001001895.1 | 14 | 2301 | ||
NM_001243467.1 | NP_001230396.1 | 12 | 2084 | |||
NM_018961.3 | NP_061834.1 | 15 | 2415 | |||
XM_006724013.3 | XP_006724076.1 | 15 | 2499 | |||
XM_011529606.2 | XP_011527908.1 | 15 | 2384 | |||
XM_011529607.2 | XP_011527909.1 | 15 | 2468 | |||
XM_011529610.2 | XP_011527912.1 | 11 | 4414 | |||
GenBank mRNA | AJ277750.1 | - | 15 | 2362 |