Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320537.1 | NP_001307466.1 | 7, 8 | 1610 | ||
NM_018964.3 | NP_061837.3 | 8, 9 | 979 | |||
XM_011529614.2 | XP_011527916.1 | 6, 7 | 1751 | |||
XM_011529615.1 | XP_011527917.1 | 6, 7 | 1751 | |||
XM_017028376.1 | XP_016883865.1 | 6, 7 | 1751 | |||
XM_017028377.1 | XP_016883866.1 | 8, 9 | 904 | |||
XM_017028378.1 | XP_016883867.1 | 8, 9 | 1363 | |||
XM_017028379.1 | XP_016883868.1 | 9, 10 | 1010 | |||
XM_017028380.1 | XP_016883869.1 | 7, 8 | 810 | |||
XM_017028381.1 | XP_016883870.1 | 8, 9 | 904 | |||
XM_017028382.1 | XP_016883871.1 | 6, 7 | 2092 | |||
XM_017028383.1 | XP_016883872.1 | 8, 9 | 835 | |||
GenBank mRNA | AF311320.1 | - | 7, 8 | 1549 | ||
AJ269529.2 | - | 8, 9 | 979 |