Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018725.3 | NP_061195.2 | 11 | 997 | ||
XM_005265310.4 | XP_005265367.1 | 12 | 1084 | |||
XM_005265311.4 | XP_005265368.1 | 11 | 1061 | |||
XM_005265312.4 | XP_005265369.1 | 10 | 1005 | |||
XM_017006804.1 | XP_016862293.1 | 11 | 953 | |||
XM_017006805.1 | XP_016862294.1 | 10 | 905 | |||
XM_017006806.1 | XP_016862295.1 | 10 | 866 | |||
GenBank mRNA | AF208110.1 | - | 11 | 963 | ||
AF208111.1 | - | 10 | 1992 | |||
AF212365.1 | - | 11 | 967 | |||
AF250309.1 | - | 11 | 958 | |||
AK307749.1 | - | 10 | 955 | |||
AY359122.1 | - | 11 | 964 | |||
BC000980.1 | - | 11 | 967 | 2985728 |