Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194954.1 | NP_001181883.1 | 17 | 3239 | ||
NM_001194955.1 | NP_001181884.1 | 15 | 2999 | |||
NM_001194956.1 | NP_001181885.1 | 14 | 1910 | |||
NM_001282278.1 | NP_001269207.1 | 17 | 2232 | |||
NM_018834.5 | NP_061322.2 | 15 | 2882 | |||
NM_199189.2 | NP_954659.1 | 18 | 3321 | |||
GenBank mRNA | AB018266.1 | - | 15 | 2753 | ||
AF117236.1 | - | 14 | 2800 | |||
AK001388.1 | - | 14 | 1702 | |||
AK293952.1 | - | 17 | 1882 | |||
AK299399.1 | - | 14 | 1886 | |||
AK307872.1 | - | 17 | 3238 | |||
AK309195.1 | - | 11 | 1996 | |||
AK316420.1 | - | 14 | 1910 | |||
AY007157.1 | - | 8 | 1234 | |||
BC015031.2 | - | 14 | 2722 | 3921668 | ||
BX537365.1 | - | 15 | 2786 | |||
BX647153.1 | - | 13 | 6246 | |||
CR749210.1 | - | 6 | 1087 | |||
CR749625.1 | - | 18 | 2972 | |||
CR936615.1 | - | 11 | 5092 | |||
M63483.1 | - | 9 | 1261 |