Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105376805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135056.1 | 2 | 400 | |||
NR_135057.1 | 2 | 397 | ||||
NR_135058.1 | 2 | 256 | ||||
NR_135059.1 | 2 | 253 | ||||
GenBank mRNA | BM923306.1 | - | 2 | 193 | ||
BX115769.1 | - | 2 | 228 | |||
HY134640.1 | - | 2 | 397 |