Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM229A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167676.1 | NP_001161148.1 | 3 | 629 | ||
GenBank mRNA | AK093734.1 | - | 2 | 2024 | ||
AK125095.1 | - | 2 | 2690 | |||
BC111382.1 | - | 2 | 471 |