Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033872.1 | 11 | 1201 | |||
GenBank mRNA | AK308634.1 | - | 3 | 296 | ||
BC040598.1 | - | 11 | 1201 |