Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018968.3 | NP_061841.2 | 17 | 1759 | ||
XM_011510366.2 | XP_011508668.1 | 15 | 1796 | |||
XM_017004361.1 | XP_016859850.1 | 17 | 1721 | |||
XM_017004363.1 | XP_016859852.1 | 17 | 1908 | |||
XM_017004364.1 | XP_016859853.1 | 14 | 5049 | |||
XM_017004367.1 | XP_016859856.1 | 18 | 1797 | |||
XM_017004373.1 | XP_016859862.1 | 12 | 2363 | |||
GenBank mRNA | AJ003029.1 | - | 17 | 1761 | ||
BC125251.1 | - | 11 | 1307 | 40025232 | ||
BG208326.1 | - | 3 | 345 |