Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319303.1 | NP_001306232.1 | 6 | 2763 | ||
NR_135067.1 | 5 | 2146 | ||||
NR_135068.1 | 2 | 1095 | ||||
XM_017016714.1 | XP_016872203.1 | 5 | 1787 | |||
XM_017016715.1 | XP_016872204.1 | 5 | 1764 | |||
GenBank mRNA | AF333487.1 | - | 3 | 888 | ||
AK172864.1 | - | 7 | 2674 | |||
AY014271.1 | - | 3 | 888 | |||
BC012394.1 | - | 2 | 995 |