Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143909.1 | NP_001137381.1 | 1 | 882 | ||
GenBank mRNA | BC137242.1 | - | 1 | 821 | 9021239 | |
BC137243.1 | - | 1 | 821 | 9021240 | ||
BC157829.1 | - | 1 | 821 | 9057048 | ||
BC157875.1 | - | 1 | 883 | 9057093 | ||
BC171806.1 | - | 1 | 821 | 9054460 | ||
BC171885.1 | - | 1 | 884 | 9054539 |