Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513013.2 | XP_011511315.2 | 10 | 4940 | ||
XM_011513019.2 | XP_011511321.2 | 9 | 2300 | |||
XM_011513020.2 | XP_011511322.2 | 10 | 2363 | |||
XM_017006898.1 | XP_016862387.1 | 11 | 3179 | |||
XM_017006899.1 | XP_016862388.1 | 11 | 5003 | |||
XM_017006900.1 | XP_016862389.1 | 10 | 3076 | |||
XM_017006901.1 | XP_016862390.1 | 11 | 4813 | |||
XM_017006902.1 | XP_016862391.1 | 11 | 3139 | |||
XM_017006905.1 | XP_016862394.1 | 12 | 4396 | |||
XM_017006906.1 | XP_016862395.1 | 12 | 4177 | |||
XM_017006907.1 | XP_016862396.1 | 12 | 3942 | |||
XM_017006908.1 | XP_016862397.1 | 12 | 4067 | |||
XM_017006909.1 | XP_016862398.1 | 11 | 3854 | |||
XM_017006912.1 | XP_016862401.1 | 10 | 3595 | |||
XM_017006914.1 | XP_016862403.1 | 10 | 3802 | |||
XM_017006915.1 | XP_016862404.1 | 10 | 3784 |