Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144952.1 | NP_001138424.1 | 12, 13 | 1591 | ||
XM_011524914.2 | XP_011523216.1 | 12, 13 | 1975 | |||
XM_011524915.2 | XP_011523217.1 | 12, 13 | 1977 | |||
XM_011524916.2 | XP_011523218.1 | 12, 13 | 1985 | |||
GenBank mRNA | AB040947.1 | - | 5, 6 | 622 | ||
BC066363.1 | - | 4, 5 | 491 |