Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286238.1 | NP_001273167.1 | 2 | 295 | ||
NM_001286239.1 | NP_001273168.1 | 2 | 295 | |||
NM_019073.3 | NP_061946.1 | 2 | 295 | |||
XM_006710699.2 | XP_006710762.1 | 2 | 289 | |||
XM_006710700.2 | XP_006710763.1 | 2 | 261 | |||
XM_006710701.3 | XP_006710764.1 | 2 | 289 | |||
XM_011541606.1 | XP_011539908.1 | 2 | 289 | |||
XM_011541608.1 | XP_011539910.1 | 2 | 281 | |||
XM_017001509.1 | XP_016856998.1 | 2 | 289 | |||
XM_017001510.1 | XP_016856999.1 | 2 | 260 | |||
XM_017001511.1 | XP_016857000.1 | 2 | 289 | |||
XM_017001512.1 | XP_016857001.1 | 2 | 93 | |||
XM_017001514.1 | XP_016857003.1 | 2 | 116 | |||
XM_017001515.1 | XP_016857004.1 | 2 | 101 | |||
GenBank mRNA | AK000869.1 | - | 2 | 260 | ||
AK301770.1 | - | 2 | 238 | |||
BC020660.1 | - | 2 | 97 | 4657425 |