Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L4A-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027706.1 | 1 | 1325 | |||
GenBank mRNA | AK002097.1 | - | 1 | 1325 | ||
BX089466.1 | - | 1 | 571 |