Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 4 | 468 | ||
XM_011544605.2 | XP_011542907.1 | 4 | 353 | |||
XM_011544606.2 | XP_011542908.1 | 4 | 404 | |||
XM_011544607.2 | XP_011542909.1 | 4 | 366 | |||
XM_017013718.1 | XP_016869207.1 | 4 | 366 | |||
XM_017013719.1 | XP_016869208.1 | 3 | 189 | |||
GenBank mRNA | AF286164.1 | - | 4 | 393 | ||
AK098787.1 | - | 4 | 469 | |||
AK291842.1 | - | 4 | 412 | |||
AK296068.1 | - | 3 | 261 |