Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012293.2 | NP_036425.1 | 19 | 3883 | ||
XM_005264707.3 | XP_005264764.1 | 18 | 3799 | |||
XM_011510396.1 | XP_011508698.1 | 19 | 3640 | |||
XM_011510397.2 | XP_011508699.1 | 18 | 3728 | |||
GenBank mRNA | AF200348.1 | - | 19 | 3842 | ||
BC070119.1 | - | 11 | 2878 | |||
D86983.1 | - | 19 | 3842 | |||
EF090903.1 | - | 19 | 3842 |