Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 15 | 1379 | ||
XM_011511359.1 | XP_011509661.1 | 15 | 1376 | |||
XM_011511361.1 | XP_011509663.1 | 14 | 1133 | |||
XM_011511362.1 | XP_011509664.1 | 14 | 1247 | |||
GenBank mRNA | AB209993.1 | - | 15 | 1298 | ||
AK001858.1 | - | 3 | 173 | |||
AK025611.1 | - | 14 | 1147 | |||
AK057926.1 | - | 15 | 1240 | |||
AK290722.1 | - | 15 | 1286 | |||
AK309040.1 | - | 15 | 1286 | |||
BC028609.1 | - | 15 | 1352 | 4826022 | ||
BC093018.1 | - | 15 | 1269 | 30337330 |