Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIM66 SIRNA Assays ›
Gene Symbol:
TRIM66
Gene Name:
tripartite motif containing 66
Gene Aliases:
C11orf29, TIF1D, TIF1DELTA
Chromosome Location:
Chr.11: 8612037 - 8683217 on Build GRCh38
UniGene ID:
Hs.130836
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
279049
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIM66
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014818.1
NP_055633.1
11
2251
XM_006718397.2
XP_006718460.1
16
3096
XM_006718398.3
XP_006718461.1
12
3708
XM_011520504.1
XP_011518806.1
17
3139
XM_011520507.2
XP_011518809.1
16
3060
XM_011520508.1
XP_011518810.1
16
3096
XM_011520509.1
XP_011518811.1
16
3096
XM_011520510.1
XP_011518812.1
16
3096
XM_011520511.1
XP_011518813.1
16
2985
XM_011520512.1
XP_011518814.1
12
2360
XM_011520513.1
XP_011518815.1
12
2358
XM_011520514.2
XP_011518816.1
12
2314
XM_011520515.1
XP_011518817.1
13
2527
XM_011520516.1
XP_011518818.1
13
2527
XM_011520517.2
XP_011518819.1
13
3849
XM_011520518.1
XP_011518820.1
13
2497
XM_011520519.1
XP_011518821.1
12
2385
XM_011520522.2
XP_011518824.1
11
3563
XM_011520523.2
XP_011518825.1
12
4445
XM_011520524.1
XP_011518826.1
16
3096
XM_011520525.1
XP_011518827.1
16
3096
XM_011520526.2
XP_011518828.1
4
1414
XM_011520527.1
XP_011518829.1
4
682
XM_017018629.1
XP_016874118.1
16
3489
XM_017018630.1
XP_016874119.1
11
2451
XM_017018631.1
XP_016874120.1
11
2603
GenBank mRNA
AB002296.2
-
11
2251
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0