Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIM66 SIRNA Assays ›
Gene Symbol:
TRIM66
Gene Name:
tripartite motif containing 66
Gene Aliases:
C11orf29, TIF1D, TIF1DELTA
Chromosome Location:
Chr.11: 8612037 - 8683217 on Build GRCh38
UniGene ID:
Hs.130836
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
279050
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIM66
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014818.1
NP_055633.1
12, 13
2687
XM_006718397.2
XP_006718460.1
17, 18
3625
XM_006718398.3
XP_006718461.1
13, 14
4237
XM_011520504.1
XP_011518806.1
18, 19
3668
XM_011520507.2
XP_011518809.1
17, 18
3589
XM_011520508.1
XP_011518810.1
17, 18
3625
XM_011520509.1
XP_011518811.1
17, 18
3625
XM_011520510.1
XP_011518812.1
17, 18
3625
XM_011520511.1
XP_011518813.1
17, 18
3514
XM_011520512.1
XP_011518814.1
13, 14
2889
XM_011520513.1
XP_011518815.1
13, 14
2887
XM_011520514.2
XP_011518816.1
13, 14
2843
XM_011520515.1
XP_011518817.1
14, 15
3056
XM_011520516.1
XP_011518818.1
14, 15
3056
XM_011520517.2
XP_011518819.1
14, 15
4378
XM_011520518.1
XP_011518820.1
14, 15
3026
XM_011520519.1
XP_011518821.1
13, 14
2914
XM_011520522.2
XP_011518824.1
12, 13
4092
XM_011520523.2
XP_011518825.1
13, 14
4974
XM_011520524.1
XP_011518826.1
17, 18
3625
XM_011520526.2
XP_011518828.1
5, 6
1943
XM_011520527.1
XP_011518829.1
5, 6
1211
XM_017018629.1
XP_016874118.1
17, 18
4018
XM_017018630.1
XP_016874119.1
12, 13
2980
XM_017018631.1
XP_016874120.1
12, 13
3132
GenBank mRNA
AB002296.2
-
11, 12
2941
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0