Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270782.1 | NP_001257711.1 | 4 | 773 | ||
NM_003201.2 | NP_003192.1 | 4 | 773 | |||
NR_073073.1 | 5 | 840 | ||||
XM_011540120.2 | XP_011538422.1 | 4 | 506 | |||
XM_011540121.2 | XP_011538423.1 | 4 | 506 | |||
GenBank mRNA | AB451241.1 | - | 4 | 368 | ||
AB451366.1 | - | 4 | 368 | |||
AK312558.1 | - | 4 | 511 | |||
BC126366.1 | - | 4 | 456 | 8992082 | ||
BM455062.1 | - | 4 | 482 | |||
BT019658.1 | - | 4 | 368 | |||
BT019659.1 | - | 4 | 368 | |||
CR407653.1 | - | 4 | 368 | |||
EU279428.1 | - | 4 | 396 | |||
M62810.1 | - | 4 | 500 |
Gene Symbol: TFAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001288.1 | 1 | 489 | |||
GenBank mRNA | CV358951.1 | - | 1 | 113 |