Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002607.5 | NP_002598.4 | 7 | 2003 | ||
NM_033023.4 | NP_148983.1 | 6 | 1934 | |||
XM_011515415.1 | XP_011513717.1 | 8 | 1495 | |||
XM_011515416.1 | XP_011513718.1 | 8 | 1499 | |||
XM_011515417.1 | XP_011513719.1 | 7 | 1426 | |||
XM_011515418.1 | XP_011513720.1 | 7 | 1430 | |||
XM_011515419.2 | XP_011513721.1 | 7 | 1203 | |||
XM_017012289.1 | XP_016867778.1 | 7 | 1236 | |||
GenBank mRNA | X06374.1 | - | 6 | 1493 |