Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002273.2 | NP_001002273.1 | 7 | 1265 | ||
NM_001002274.2 | NP_001002274.1 | 7 | 1268 | |||
NM_001002275.2 | NP_001002275.1 | 8 | 1322 | |||
NM_001190828.1 | NP_001177757.1 | 7 | 1304 | |||
NM_004001.4 | NP_003992.3 | 8 | 1325 | |||
XM_017000671.1 | XP_016856160.1 | 9 | 1289 | |||
XM_017000672.1 | XP_016856161.1 | 8 | 1232 | |||
GenBank mRNA | AB209585.1 | - | 6 | 4446 | ||
AF543826.1 | - | 8 | 1278 | |||
AK308150.1 | - | 7 | 1195 | |||
BC031992.1 | - | 8 | 1235 | 4752609 | ||
BC141826.1 | - | 7 | 1178 | 40148276 | ||
BC146678.1 | - | 7 | 1183 | 40148906 | ||
H04104.1 | - | 1 | 60 | |||
M28696.1 | - | 7 | 1181 | |||
M31933.1 | - | 7 | 1254 | |||
M31934.1 | - | 7 | 1177 | |||
M31935.1 | - | 8 | 1145 | |||
X17653.1 | - | 7 | 1210 | |||
X52473.1 | - | 7 | 1186 |