Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182505.3 | NP_872311.2 | 4 | 687 | ||
XM_011518226.2 | XP_011516528.1 | 5 | 985 | |||
GenBank mRNA | BC010179.2 | - | 3 | 576 | ||
BC052375.1 | - | 4 | 526 | 6147448 | ||
BQ431408.1 | - | 4 | 488 | |||
CB113688.1 | - | 2 | 183 |