Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099625.1 | NP_001093095.1 | 7 | 1067 | ||
NM_001099626.1 | NP_001093096.1 | 7 | 1119 | |||
NM_001099627.1 | NP_001093097.1 | 7 | 958 | |||
NM_019557.5 | NP_062457.3 | 7 | 1165 | |||
GenBank mRNA | AB075880.1 | - | 7 | 995 | ||
AF173891.1 | - | 7 | 932 | |||
AK056721.1 | - | 6 | 2321 | |||
AK127164.1 | - | 3 | 1601 | |||
AK290744.1 | - | 6 | 754 | |||
AK299221.1 | - | 7 | 1010 | |||
AK314297.1 | - | 7 | 995 | |||
AL512766.1 | - | 7 | 1167 | |||
AY007100.1 | - | 7 | 1045 | |||
BC017175.2 | - | 7 | 1119 | 3162883 | ||
BX647921.1 | - | 7 | 915 | |||
KU178723.1 | - | 6 | 857 | |||
KU178724.1 | - | 6 | 880 |