Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032172.2 | NP_115548.1 | 18 | 4125 | ||
XM_006715790.1 | XP_006715853.1 | 17 | 4028 | |||
XM_006715791.2 | XP_006715854.1 | 20 | 4534 | |||
XM_011515573.1 | XP_011513875.1 | 21 | 4593 | |||
XM_011515574.1 | XP_011513876.1 | 18 | 4117 | |||
XM_011515577.1 | XP_011513879.1 | 16 | 3746 | |||
XM_011515578.1 | XP_011513880.1 | 18 | 4168 | |||
GenBank mRNA | AK309375.1 | - | 15 | 1712 | ||
BC132862.1 | - | 17 | 4022 | 40146884 | ||
BC136656.1 | - | 16 | 4955 | |||
BF725643.1 | - | 3 | 222 | |||
BU187025.1 | - | 4 | 512 |