Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024096.1 | 2 | 987 | |||
GenBank mRNA | AK094097.1 | - | 2 | 1400 | ||
BC132989.1 | - | 2 | 738 | |||
BC132991.1 | - | 2 | 738 | |||
BX161445.1 | - | 2 | 987 |