Gene Symbol: NHLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301159.1 | NP_001288088.1 | 2 | 1844 | ||
NM_176677.2 | NP_788850.1 | 2 | 1819 | |||
GenBank mRNA | AK093527.1 | - | 2 | 1802 | ||
BC032015.1 | - | 2 | 1782 | |||
BC048340.1 | - | 2 | 1781 |